3,6-Dibromo-9-phenylcarbazole 98%

Supplier: Ambeed
Warning

A105131-100G A105131-10G A105131-1G A105131-5G A105131-25G
77270-450EA 79 USD
77270-450 77270-452 77270-454 77270-456 77747-783
3,6-Dibromo-9-phenylcarbazole 98%
3,6-Dibromo-9-phenylcarbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₁Br₂N
MDL Number: MFCD08276338
CAS Number: 57103-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR