4-Oxo-3,4-dihydro-phthalazine-1-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-Oxo-3,4-dihydrophthalazine-1-carboxylic acid

A105122-100MG A105122-250MG A105122-1G A105122-5G
77283-340EA 32.96 USD
77283-340 77283-342 77570-304 77747-782
4-Oxo-3,4-dihydro-phthalazine-1-carboxylic acid 95%
4-Oxo-3,4-dihydro-phthalazine-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆N₂O₃
MW: 190.156 g/mol
MDL Number: MFCD00524727
CAS Number: 3260-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR