3-Chloro-o-cresol 97%

Supplier: Ambeed
Warning

A105102-100G A105102-10G A105102-1G A105102-25G A105102-5G
77270-440EA 136 USD
77270-440 77270-442 77270-444 77270-446 77270-448
3-Chloro-o-cresol 97%
3-Chloro-o-cresol

Precaución: for Research Use Only

Formula: C₇H₇ClO
MDL Number: MFCD00079738
CAS Number: 3260-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR