(R)-3,3'-Bis3,5-bis(trifluoromethyl)phenyl)-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2,2'-naphthol 98%
Supplier: Ambeed
|
Warning
|
A105093-100MG
A105093-1G
A105093-250MG
A105093-5G
77245-360EA
62.83
USD
77245-360
77245-362
77245-364
77245-366
(R)-3,3'-Bis3,5-bis(trifluoromethyl)phenyl)-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2,2'-naphthol 98%
(R)-3,3'-Bis3,5-bis(trifluoromethyl)phenyl)-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2,2'-naphthol
Precaución: for Research Use Only
Formula:
C₃₆H₂₆F₁₂O₂ MW: 718.57 g/mol |
MDL Number:
MFCD31560041 CAS Number: 618854-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...