2-(3-Methylbut-2-enamido)acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Methylcrotonylglycine

A105022-100MG A105022-250MG
77245-356EA 86.52 USD
77245-356 77245-358
2-(3-Methylbut-2-enamido)acetic acid 95%
2-(3-Methylbut-2-enamido)acetic acid

Precaución: for Research Use Only

Formula: C₇H₁₁NO₃
MW: 157.17 g/mol
MDL Number: MFCD16293865
CAS Number: 33008-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR