4-(2,6-Dichloro-4-pyrimidyl)morpholine 98%

Supplier: Ambeed
Warning

Synonyms: 4-(2,6-Dichloropyrimidin-4-yl)morpholine, 2,4-Dichloro-6-(4-morpholino)pyrimidine

A104986-250MG A104986-1G A104986-5G
77283-314EA 33.99 USD
77283-314 77570-294 77747-781
4-(2,6-Dichloro-4-pyrimidyl)morpholine 98%
4-(2,6-Dichloro-4-pyrimidyl)morpholine

Precaución: for Research Use Only

Formula: C₈H₉Cl₂N₃O
MW: 234.08 g/mol
MDL Number: MFCD09746259
CAS Number: 52127-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR