2-Amino-6-methylpyridin-3-ol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-hydroxy-6-methylpyridine

A104964-1G A104964-250MG A104964-5G
77257-402EA 47 USD
77257-402 77257-404 77257-406
2-Amino-6-methylpyridin-3-ol 97%
2-Amino-6-methylpyridin-3-ol

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MW: 124.1400 g/mol
MDL Number: MFCD09839245
CAS Number: 20348-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR