2,7-Dibromo-9,9-dimethyl-9,10-dihydroacridine 97%

Supplier: Ambeed
Warning

A104946-100MG A104946-10G A104946-1G A104946-250MG A104946-25G A104946-5G
77233-194EA 25 USD
77233-194 77233-196 77233-198 77233-200 77233-202 77233-204
2,7-Dibromo-9,9-dimethyl-9,10-dihydroacridine 97%
2,7-Dibromo-9,9-dimethyl-9,10-dihydroacridine

Precaución: for Research Use Only

Formula: C₁₅H₁₃Br₂N
MDL Number: MFCD26402967
CAS Number: 1333316-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR