Methyl 2,6-dimethyl-4-oxocyclohex-2-enecarboxylate 97%

Supplier: Ambeed
Warning

A104853-10G A104853-5G
77233-190EA 12.36 USD
77233-190 77233-192
Methyl 2,6-dimethyl-4-oxocyclohex-2-enecarboxylate 97%
Methyl 2,6-dimethyl-4-oxocyclohex-2-enecarboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃
MW: 182.22 g/mol
MDL Number: MFCD22571578
CAS Number: 67333-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR