4-(Aminomethyl)cyclohexanecarboxylic acid (cis and trans mixture) 95%

Supplier: Ambeed
Warning

A104837-25G A104837-5G
77283-270EA 55.62 USD
77283-270 77283-272 77283-274 77283-276 77283-278
4-(Aminomethyl)cyclohexanecarboxylic acid (cis and trans mixture) 95%
4-(Aminomethyl)cyclohexanecarboxylic acid (cis and trans mixture)

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂
MDL Number: MFCD00064951
CAS Number: 701-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR