3-Chlorothiophene 96%

Supplier: Ambeed
Danger

A104823-10G A104823-1G A104823-25G A104823-5G
77270-422EA 13 USD
77270-422 77270-424 77270-426 77270-428
3-Chlorothiophene 96%
3-Chlorothiophene

Precaución: for Research Use Only

Formula: C₄H₃ClS
MDL Number: MFCD00043887
CAS Number: 17249-80-8
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR