Methyl-2-amino-3-nitrobenzoate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-3-nitrobenzoate

A104712-100G A104712-1G A104712-5G A104712-10G A104712-25G
77245-334EA 154.5 USD
77245-334 77245-336 77245-338 77745-792 77745-793
Methyl-2-amino-3-nitrobenzoate 95%
Methyl 2-amino-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₈N₂O₄
MW: 196.1600 g/mol
MDL Number: MFCD02093531
CAS Number: 57113-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR