2-Amino-5-chloro-6-methylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-6-methylpyridin-2-amine

A104691-10G A104691-250MG A104691-25G A104691-5G
77245-326EA 189.52 USD
77245-326 77245-328 77245-330 77245-332
2-Amino-5-chloro-6-methylpyridine 97%
2-Amino-5-chloro-6-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MW: 142.59 g/mol
MDL Number: MFCD09453335
CAS Number: 36936-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR