1-Bromo-2,3-dichlorobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dichlorobromobenzene

A104560-100G A104560-10G A104560-25G A104560-500G
77233-174EA 52 USD
77233-174 77233-176 77233-178 77233-180
1-Bromo-2,3-dichlorobenzene 97%
1-Bromo-2,3-dichlorobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 243
Melting Pt: 60
MDL Number: MFCD00000536
CAS Number: 56961-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR