L-Isoleucine-tert-butyl ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-Ile-OtBu.HCl, tert-Butyl (2S,3S)-2-amino-3-methylpentanoate hydrochloride

A104417-1G A104417-250MG A104417-25G A104417-5G A104417-10G
77270-410EA 9 USD
77270-410 77270-412 77270-414 77270-416 77570-270
L-Isoleucine-tert-butyl ester hydrochloride 98%
L-Isoleucine-tert-butyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₂₁NO₂
MW: 223.74 g/mol
MDL Number: MFCD00058015
CAS Number: 69320-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR