6-Bromo-1H-indene 95% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A104326-1G A104326-250MG A104326-5G
77233-170EA 105.06 USD
77233-170 77233-172 77570-262
6-Bromo-1H-indene 95% stabilized
6-Bromo-1H-indene

Precaución: for Research Use Only

Formula: C₉H₇Br
MW: 195.05 g/mol
MDL Number: MFCD08234616
CAS Number: 33065-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR