2,6-Diamino-3,5-difluoropyridine 98%

Supplier: Ambeed
Warning

A104324-1G A104324-25G
77245-306EA 14.42 USD
77245-306 77245-308
2,6-Diamino-3,5-difluoropyridine 98%
2,6-Diamino-3,5-difluoropyridine

Precaución: for Research Use Only

Formula: C₅H₅F₂N₃
MW: 145.11 g/mol
MDL Number: MFCD03412208
CAS Number: 247069-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR