6-Fluoro-7-nitro-1H-indazole 97%

Supplier: Ambeed
Warning

A104291-100MG A104291-1G A104291-250MG A104291-5G
77270-402EA 18 USD
77270-402 77270-404 77270-406 77270-408
6-Fluoro-7-nitro-1H-indazole 97%
6-Fluoro-7-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄FN₃O₂
MW: 181.12 g/mol
MDL Number: MFCD31619104
CAS Number: 1279869-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR