VUF 10460 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Methylpiperazin-1-yl)-6-phenylpyrimidin-2-amine, VUF10460

A104244-10MG A104244-1MG A104244-25MG A104244-50MG A104244-5MG
77270-392EA 89 USD
77270-392 77270-394 77270-396 77270-398 77270-400
VUF 10460 98%
VUF 10460

Precaución: for Research Use Only

Formula: C₁₅H₁₉N₅
MW: 269.34 g/mol
MDL Number: MFCD13187037
CAS Number: 1028327-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR