3-(Piperidinosulfonyl)benzenecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Piperidin-1-ylsulphonyl)benzoic acid

A104242-1G A104242-250MG A104242-5G
77270-386EA 44.29 USD
77270-386 77270-388 77270-390
3-(Piperidinosulfonyl)benzenecarboxylic acid 97%
3-(Piperidinosulfonyl)benzenecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₄S
MW: 269.32 g/mol
MDL Number: MFCD00760901
CAS Number: 7311-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR