Thiosalicylic acid 98%

Supplier: Ambeed
Danger

Synonyms: 2-Thiobenzoic acid, 2-Mercaptobenzoic acid

A104228-100G A104228-25G A104228-500G A104228-5G
77257-374EA 20.6 USD
77257-374 77257-376 77257-378 77257-380
Thiosalicylic acid 98%
Thiosalicylic acid

Precaución: for Research Use Only

Formula: C₇H₆O₂S
MW: 154.19 g/mol
MDL Number: MFCD00004836
CAS Number: 147-93-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR