1,4-Difluoro-2-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Difluoronitrobenzene

A103936-1000G A103936-100G A103936-10G A103936-25G A103936-500G A103936-5G
77245-268EA 204 USD
77245-268 77245-270 77245-272 77245-274 77245-276 77245-278
1,4-Difluoro-2-nitrobenzene 98%
1,4-Difluoro-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 207
Melting Pt: -11.7
MDL Number: MFCD00007054
CAS Number: 364-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR