4-Aminosalicylic acid 95%

Supplier: Ambeed
Warning

A103805-25G A103805-500G A103805-100G A103805-1000G
77283-162EA 12 USD
77283-162 77283-164 77565-798 77747-776
4-Aminosalicylic acid 95%
4-Aminosalicylic acid

Precaución: For research use only.

Formula: C₇H₇NO₃
MDL Number: MFCD00007789
CAS Number: 65-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR