1,2,4,5-Tetrabromo-3,6-dichlorobenzene 97%

Supplier: Ambeed
Warning

A103731-1G A103731-250MG
77245-264EA 221.45 USD
77245-264 77245-266
1,2,4,5-Tetrabromo-3,6-dichlorobenzene 97%
1,2,4,5-Tetrabromo-3,6-dichlorobenzene

Precaución: for Research Use Only

Formula: C₆Br₄Cl₂
MW: 462.59 g/mol
MDL Number: MFCD00192661
CAS Number: 31604-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR