2,9-Dibutyl-1,10-phenanthroline 98%

Supplier: Ambeed
Warning

A103709-250MG A103709-1G A103709-100MG A103709-5G
77245-262EA 39.14 USD
77245-262 77548-960 77614-778 77745-790
2,9-Dibutyl-1,10-phenanthroline 98%
2,9-Dibutyl-1,10-phenanthroline

Precaución: for Research Use Only

Formula: C₂₀H₂₄N₂
MW: 292.43 g/mol
MDL Number: MFCD02093459
CAS Number: 85575-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR