2-(4-Ethylphenyl)-2-methylpropanoic acid 98%

Supplier: Ambeed
Warning

A103637-100G A103637-25G
77245-258EA 124 USD
77245-258 77245-260
2-(4-Ethylphenyl)-2-methylpropanoic acid 98%
2-(4-Ethylphenyl)-2-methylpropanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
MDL Number: MFCD13180614
CAS Number: 1247119-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR