3-(2-Cyanophenyl)propanoic acid 98%

Supplier: Ambeed
Warning

A103626-1G A103626-250MG A103626-5G
77270-362EA 91 USD
77270-362 77270-364 77270-366
3-(2-Cyanophenyl)propanoic acid 98%
3-(2-Cyanophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MDL Number: MFCD00464857
CAS Number: 27916-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR