2,4-Dichloro-1-(4-chloro-2-methoxyphenoxy)benzene 97%

Supplier: Ambeed
Warning

Synonyms: Triclosan-methyl

A103616-1G A103616-250MG
77245-248EA 97.85 USD
77245-248 77245-250
2,4-Dichloro-1-(4-chloro-2-methoxyphenoxy)benzene 97%
2,4-Dichloro-1-(4-chloro-2-methoxyphenoxy)benzene

Precaución: for Research Use Only

Formula: C₁₃H₉Cl₃O₂
MW: 303.56 g/mol
MDL Number: MFCD04039700
CAS Number: 4640-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR