2-Fluoro-4-(trans-4-pentylcyclohexyl)-4'-(trans-4-propylcyclohexyl)biphenyl 97%

Supplier: Ambeed
Warning

A103603-1G A103603-5G
77283-132EA 14 USD
77283-132 77283-134
2-Fluoro-4-(trans-4-pentylcyclohexyl)-4'-(trans-4-propylcyclohexyl)biphenyl 97%
2-Fluoro-4-(trans-4-pentylcyclohexyl)-4'-(trans-4-propylcyclohexyl)biphenyl

Precaución: for Research Use Only

Formula: C₃₂H₄₅F
MDL Number: MFCD11053473
CAS Number: 106349-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR