5-Chlorouracil 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloropyrimidine-2,4(1H,3H)-dione, Chlorouracil

A103527-100G A103527-1G A103527-25G
77270-352EA 55.62 USD
77270-352 77270-354 77270-356
5-Chlorouracil 98%
5-Chlorouracil

Precaución: for Research Use Only

Formula: C₄H₃ClN₂O₂
MW: 146.53 g/mol
MDL Number: MFCD00006019
CAS Number: 1820-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR