2-(2,6-Dichlorophenyl)ethanol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,6-Dichlorophenethyl Alcohol
A103463-100G
A103463-10G
A103463-1G
A103463-25G
A103463-500G
A103463-5G
77245-228EA
231.01
USD
77245-228
77245-230
77245-232
77245-234
77245-236
77245-238
77245-240
2-(2,6-Dichlorophenyl)ethanol 98%
2-(2,6-Dichlorophenyl)ethanol
Precaución: for Research Use Only
Formula:
C₈H₈Cl₂O |
MDL Number:
MFCD00800674 CAS Number: 30595-79-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...