3-Methyl-4-nitro-1H-pyrazole 97%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-4-nitropyrazole

A103301-100G A103301-10G A103301-1G A103301-250MG A103301-25G A103301-500G A103301-5G
77270-336EA 58.71 USD
77270-336 77270-338 77270-340 77270-342 77270-344 77270-346 77570-194
3-Methyl-4-nitro-1H-pyrazole 97%
3-Methyl-4-nitro-1H-pyrazole

Precaución: for Research Use Only

Formula: C4H5N3O2
MW: 127.10 g/mol
MDL Number: MFCD00037864
CAS Number: 5334-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR