2-Chloropyrimidine 98%

Supplier: Ambeed
Warning

A103237-1000G A103237-100G A103237-10G A103237-25G A103237-500G A103237-5G A103237-1G
77283-058EA 297.67 USD
77283-058 77283-060 77283-062 77283-064 77283-066 77283-068 77570-188
2-Chloropyrimidine 98%
2-Chloropyrimidine

Precaución: for Research Use Only

Formula: C₄H₃ClN₂
MW: 114.53 g/mol
MDL Number: MFCD00006060
CAS Number: 1722-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR