IDF-11774 98+%

Supplier: Ambeed
Warning

A103226-10MG A103226-1MG A103226-5MG A103226-50MG
77257-326EA 47 USD
77257-326 77257-328 77257-330 77747-767
IDF-11774 98+%
IDF-11774

Precaución: for Research Use Only

Formula: C₂₃H₃₂N₂O₂
MDL Number: MFCD31619285
CAS Number: 1429054-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR