6-Fluoro-1H-indole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoroindole-2-carboxylic acid

A103139-10G A103139-1G A103139-250MG A103139-25G A103139-5G
77270-316EA 265.74 USD
77270-316 77270-318 77270-320 77270-322 77270-324
6-Fluoro-1H-indole-2-carboxylic acid 97%
6-Fluoro-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆FNO₂
MW: 179.15 g/mol
MDL Number: MFCD01863162
CAS Number: 3093-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR