2-Chloro-3-cyanopyrazine 98%

Supplier: Ambeed
Warning

Synonyms: 3-chloropyrazine-2-carbonitrile

A103125-100G A103125-10G A103125-25G A103125-5G
77283-036EA 99.91 USD
77283-036 77283-038 77283-040 77283-042
2-Chloro-3-cyanopyrazine 98%
2-Chloro-3-cyanopyrazine

Precaución: for Research Use Only

Formula: C₅H₂ClN₃
MW: 139.5400 g/mol
MDL Number: MFCD00219653
CAS Number: 55557-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR