2-Amino-4-fluoro-3-methylbenzoic acid 97%

Supplier: Ambeed
Warning

A102964-100MG A102964-250MG A102964-5G A102964-1G
77245-216EA 20.6 USD
77245-216 77245-218 77245-220 77565-778
2-Amino-4-fluoro-3-methylbenzoic acid 97%
2-Amino-4-fluoro-3-methylbenzoic acid

Precaución: For research use only.

Formula: C₈H₈FNO₂
MW: 169.15 g/mol
MDL Number: MFCD01318256
CAS Number: 129833-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR