6-((E)-2-(3-((E)-2-Methoxy-4-(pyridin-2-ylmethoxy)styryl)-1H-pyrazol-5-yl)vinyl)-1H-indole 97%

Supplier: Ambeed
Warning

Synonyms: PE859

A102932-10MG A102932-1MG A102932-25MG A102932-50MG A102932-5MG
77233-142EA 60.77 USD
77233-142 77233-144 77233-146 77233-148 77233-150
6-((E)-2-(3-((E)-2-Methoxy-4-(pyridin-2-ylmethoxy)styryl)-1H-pyrazol-5-yl)vinyl)-1H-indole 97%
6-((E)-2-(3-((E)-2-Methoxy-4-(pyridin-2-ylmethoxy)styryl)-1H-pyrazol-5-yl)vinyl)-1H-indole

Precaución: for Research Use Only

Formula: C₂₈H₂₄N₄O₂
MW: 448.52 g/mol
MDL Number: MFCD28411710
CAS Number: 1402727-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR