Licarin B ≥99%

Supplier: Ambeed
Warning

Synonyms: 5-((2R,3R)-7-Methoxy-3-methyl-5-((E)-prop-1-en-1-yl)-2,3-dihydrobenzofuran-2-yl)benzo[d][1,3]dioxole

A102913-1MG A102913-5MG A102913-10MG
77245-212EA 13.39 USD
77245-212 77245-214 77614-772
Licarin B ≥99%
Licarin B

Precaución: for Research Use Only

Formula: C₂₀H₂₀O₄
MW: 324.37 g/mol
MDL Number: MFCD13195533
CAS Number: 51020-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR