1-Bromo-2-chlorobenzene 97%
Supplier: Ambeed
|
Warning
|
A102846-1000G
A102846-100G
A102846-10G
A102846-25G
A102846-500G
77257-314EA
107.12
USD
77257-314
77257-316
77257-318
77257-320
77257-322
1-Bromo-2-chlorobenzene 97%
1-Bromo-2-chlorobenzene
Precaución: for Research Use Only
Formula:
C₆H₄BrCl |
MDL Number:
MFCD00000532 CAS Number: 694-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...