4-Aminobenzene-1,3-diol hydrochloride 96%

Supplier: Ambeed
Warning

A102834-100MG A102834-10G A102834-1G A102834-250MG A102834-25G A102834-5G
77282-996EA 7 USD
77282-996 77282-998 77283-000 77283-002 77283-004 77283-006
4-Aminobenzene-1,3-diol hydrochloride 96%
4-Aminobenzene-1,3-diol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₈ClNO₂
MDL Number: MFCD00012963
CAS Number: 34781-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR