7-Nitro-1H-indole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: CRT0044876

A102798-1G A102798-1MG A102798-250MG A102798-25MG A102798-50MG A102798-5G
77257-302EA 32.96 USD
77257-302 77257-304 77257-306 77257-308 77257-310 77257-312
7-Nitro-1H-indole-2-carboxylic acid 97%
7-Nitro-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆N₂O₄
MW: 206.16 g/mol
MDL Number: MFCD00044720
CAS Number: 6960-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR