(3,5-Dimethylisoxazol-4-yl)methylamine 97%

Supplier: Ambeed
Danger

A102755-100MG A102755-1G A102755-250MG A102755-5G
77270-300EA 58 USD
77270-300 77270-302 77270-304 77270-306
(3,5-Dimethylisoxazol-4-yl)methylamine 97%
(3,5-Dimethylisoxazol-4-yl)methylamine

Precaución: for Research Use Only

Formula: C₆H₁₀N₂O
MDL Number: MFCD06213033
CAS Number: 131052-47-6
UN: 3267
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR