2-Methyl-4-pentenoic acid 98%

Supplier: Ambeed
Danger

Synonyms: 2-Methylpent-4-enoic acid

A102744-25G A102744-5G
77270-296EA 43.26 USD
77270-296 77270-298
2-Methyl-4-pentenoic acid 98%
2-Methyl-4-pentenoic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
MDL Number: MFCD00015570
CAS Number: 1575-74-2
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR