2,4-Di([1,1'-biphenyl]-3-yl)-6-chloro-1,3,5-triazine 97%

Supplier: Ambeed
Warning

A102720-100MG A102720-1G A102720-250MG
77245-202EA 10 USD
77245-202 77245-204 77245-206
2,4-Di([1,1'-biphenyl]-3-yl)-6-chloro-1,3,5-triazine 97%
2,4-Di([1,1'-biphenyl]-3-yl)-6-chloro-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₂₇H₁₈ClN₃
MW: 419.91 g/mol
MDL Number: MFCD30536777
CAS Number: 1205748-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR