2,1,3-Benzothiadiazole 98%

Supplier: Ambeed
Warning

A102714-100G A102714-10G A102714-25G A102714-500G A102714-5G
77282-970EA 53.56 USD
77282-970 77282-972 77282-974 77282-976 77282-978
2,1,3-Benzothiadiazole 98%
2,1,3-Benzothiadiazole

Precaución: for Research Use Only

Formula: C₆H₄N₂S
MW: 136.18 g/mol
MDL Number: MFCD00005809
CAS Number: 273-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR