2-Bromoquinolin-4-amine 97%

Supplier: Ambeed
Warning

A102676-100MG A102676-1G A102676-250MG
77282-954EA 27 USD
77282-954 77282-956 77282-958
2-Bromoquinolin-4-amine 97%
2-Bromoquinolin-4-amine

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MW: 223.06 g/mol
MDL Number: MFCD08705640
CAS Number: 36825-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR