2-Fluoro-4-(trifluoromethyl)benzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: (2-Fluoro-4-(trifluoromethyl)phenyl)methanol

A102593-1G A102593-250MG A102593-25G A102593-5G
77233-132EA 18 USD
77233-132 77233-134 77233-136 77233-138
2-Fluoro-4-(trifluoromethyl)benzyl alcohol 95%
2-Fluoro-4-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆F₄O
MDL Number: MFCD00070808
CAS Number: 197239-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR