3-Methylbenzo[b]thiophene-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A102553-100G A102553-1G A102553-250MG A102553-25G A102553-5G
77270-284EA 1847.82 USD
77270-284 77270-286 77270-288 77270-290 77270-292
3-Methylbenzo[b]thiophene-2-carboxylic acid 98%
3-Methylbenzo[b]thiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈O₂S
MW: 192.24 g/mol
MDL Number: MFCD01830305
CAS Number: 3133-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR