2-Amino-2-ethyl-1,3-propanediol 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-2-ethylpropane-1,3-diol

A102491-100G A102491-500G A102491-25G
77245-188EA 22.66 USD
77245-188 77245-190 77745-781
2-Amino-2-ethyl-1,3-propanediol 95%
2-Amino-2-ethyl-1,3-propanediol

Precaución: for Research Use Only

Formula: C₅H₁₃NO₂
MW: 119.16 g/mol
MDL Number: MFCD00004680
CAS Number: 115-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR